missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
FABP5/E-FABP Antibody (FABP5/3750), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79929UV
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
FABP5/E-FABP Monoclonal specifically detects FABP5/E-FABP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Tekniske data
| FABP5/E-FABP | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| FABP5 | |
| Recombinant full-length human FABP5/E-FABP protein (Uniprot: Q01469) | |
| 0.1 mL | |
| Stem Cell Markers | |
| Human | |
| IgG2a κ |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| FABP5/3750 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| EFABP, E-FABPPAFABP, fatty acid binding protein 5 (psoriasis-associated), Fatty acid-binding protein 5, PA-FABPepidermal, Psoriasis-associated fatty acid-binding protein homolog | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion