missing translation for 'onlineSavingsMsg'
Learn More
Learn More
F4/80 Rat anti-Mouse, BV421, Clone: T45-2342, BD Horizon™
Rat Monoclonal Antibody
Brand: BD Biosciences 565411
1949.54 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
F4/80 Monoclonal antibody specifically detects F4/80 in Mouse samples. It is validated for Flow cytometry.
Specifications
F4/80 | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Rat | |
50 μg | |
Cell Biology | |
Mouse | |
Purified |
Flow Cytometry | |
T45-2342 | |
Brilliant Violet 421 | |
Gpf480; F480; Emr1; Ly71; DD7A5-7; EGF-TM7; TM7LN3 | |
Mouse F4/80 Recombinant Protein | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV421 under optimum conditions and unconjugated antibody and free BD Horizon™ BV421 were removed. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
F4/80 Rat anti-Mouse, BV421, Clone: T45-2342, BD Horizon™ > 50μg; BV421
For Reseach Use Only.
Spot an opportunity for improvement?Share a Content Correction