missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ F(ab')2-Goat anti-Rabbit IgG (H+L) Secondary Antibody, Qdot™ 525
Beskrivelse
Qdot nanocrystals are composed of semi-conductor material to generate a fluorescent particle which is exceptionally bright and does not photobleach. Qdot nanocrystals paired with the correct optical filters are as much as 50 times brighter than traditional organic dyes.
Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Tekniske data
Tekniske data
| Antigen | Rabbit IgG (H+L) |
| Applikationer | Flow Cytometry, Immunohistochemistry, Western Blot, Immunocytochemistry, Multiplex Immunohistochemistry |
| Klassifikation | Polyclonal |
| Konjugeret | Qdot 525 |
| Formulering | 0.05M borate with 1M betaine and 0.05% sodium azide; pH 8.3 |
| Værtsarter | Goat |
| Immunogen | Gamma Immunoglobins Heavy and Light chains. |
| Oprensningsmetode | Purified |
| Mængde | 200 μL |
| Regulatorisk status | RUO |
| Vis mere |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?