missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ F(ab')2-Goat anti-Human IgG, IgM, IgA (H+L) Secondary Antibody, FITC
F(ab')2-Goat Polyclonal Secondary Antibody
Brand: Invitrogen™ A24495
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
The sensitivity of each lot of antibody is confirmed using ELISA. The specificity of each lot of antibody is confirmed by immunoelectrophoresis (IEP).
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Tekniske data
| Human IgG, IgM, IgA (H+L) | |
| Polyclonal | |
| FITC | |
| Goat | |
| Affinity Chromatography | |
| RUO | |
| Human | |
| Secondary Antibody | |
| IgG |
| Flow Cytometry, Immunocytochemistry | |
| 1.5 mg/mL | |
| PBS with 1% BSA and 0.05% sodium azide; pH 7.8 | |
| Human IgG/A/M. | |
| 500 μg | |
| Secondary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
| Liquid |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion