Learn More
F(ab')2-Goat anti-Human IgD Fc Secondary Antibody, FITC, Invitrogen™
F(ab')2-Goat Polyclonal Secondary Antibody
Brand: Invitrogen H15501
Additional Details : Weight : 0.50000kg
Description
Commonly used, FITC conjugates provide relatively high absorptivity, excellent fluorescence quantum yield, and good water solubility.
Human IgD Fc Polyclonal antibody specifically detects Human IgD Fc in Human samples. It is validated for Flow Cytometry.Specifications
Human IgD Fc | |
Polyclonal | |
FITC | |
Assay-dependent | |
Goat | |
1 mL | |
Secondary | |
0.1% Sodium azide | |
4° C, store in dark | |
IgG |
Flow Cytometry | |
0.2mg/mL | |
F(ab')2-Goat anti-Human IgD Fc Secondary Antibody, 1mL, 0.2mg/mL Concentration, Liquid, Goat Host, Human Species Reactivity, FITC Conjugate, IgG Isotype, 4°C Storage Temperature, PBS with BSA Storage Buffer, For Flow Cytometry | |
PBS with BSA and 0.1% sodium azide | |
purified | |
ASR, RUO | |
1 Each | |
Human | |
Liquid | |
HRP, AP, fluorescence |
Analyte Specific Reagent. Analytical and performance characteristics are not established.