Få mere at vide
Ezetimibe Tetrahydropyran Impurity, TRC
Artikelnummer. 17997814
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17997814 50 mg
17928974 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17997814 Leverandør Toronto Research Chemicals Leverandørnr. TRCE97508050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1296129-15-1
Molekylær formel C24 H21 F2 N O3
Molekylvægt (g/mol) 409.43
Synonym 2H-Pyran-3-carboxamide, N,6-bis(4-fluorophenyl)tetrahydro-2-(4-hydroxyphenyl)-, (2R,3R,6S)-, (2R,3R,6S)-N,6-bis(4-fluorophenyl)-2-(4-hydroxyphenyl)-3,4,5,6-tetrahydro-2H-pyran-3-carboxamide
IUPAC navn (2R,3R,6S)-N,6-bis(4-fluorophenyl)-2-(4-hydroxyphenyl)oxane-3-carboxamide
SMIL Oc1ccc(cc1)[C@@H]2O[C@@H](CC[C@H]2C(=O)Nc3ccc(F)cc3)c4ccc(F)cc4
Tekniske data
Kemisk navn eller materiale Ezetimibe Tetrahydropyran Impurity
Renhedsgrad noter HPLC
Mængde 50 mg
InChI formel InChI=1S/C24H21F2NO3/c25-17-5-1-15(2-6-17)22-14-13-21(23(30-22)16-3-11-20(28)12-4-16)24(29)27-19-9-7-18(26)8-10-19/h1-12,21-23,28H,13-14H2,(H,27,29)/t21-,22+,23+/m1/s1
Formel vægt 409.1489
Procent renhed >95%