Få mere at vide
Ezetimibe Phenoxy Beta-D-Glucuronide, TRC
Artikelnummer. 17981815
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17981815 5 mg
17974835 10 mg
17912805 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17981815 Leverandør Toronto Research Chemicals Leverandørnr. TRCE9750305MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 190448-57-8
Molekylær formel C30 H29 F2 N O9
Molekylvægt (g/mol) 585.55
Synonym Ezetimibe Phenoxy Glucuronide
IUPAC navn (2S,3S,4S,5R,6S)-6-[4-[(2S,3R)-1-(4-fluorophenyl)-3-[(3S)-3-(4-fluorophenyl)-3-hydroxypropyl]-4-oxoazetidin-2-yl]phenoxy]-3,4,5-trihydroxyoxane-2-carboxylic acid
SMIL O[C@@H](CC[C@@H]1[C@H](N(C1=O)c2ccc(F)cc2)c3ccc(O[C@@H]4O[C@@H]([C@@H](O)[C@H](O)[C@H]4O)C(=O)O)cc3)c5ccc(F)cc5
Tekniske data
Kemisk navn eller materiale Ezetimibe Phenoxy beta-D-Glucuronide
Mængde 5 mg
InChI formel InChI=1S/C30H29F2NO9/c31-17-5-1-15(2-6-17)22(34)14-13-21-23(33(28(21)38)19-9-7-18(32)8-10-19)16-3-11-20(12-4-16)41-30-26(37)24(35)25(36)27(42-30)29(39)40/h1-12,21-27,30,34-37H,13-14H2,(H,39,40)/t21-,22+,23-,24+,25+,26-,27+,30-/m1/s1
Formel vægt 585.181