Få mere at vide
Ezetimibe Ketone Benzyl Ether, Mikromol™
Artikelnummer. 30050474
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30050474 100 mg
30050292 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30050474 Leverandør Mikromol Leverandørnr. MM0935.03
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Ezetimibe Ketone Benzyl Ether
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ezetimibe Ketone Benzyl Ether
CAS 190595-65-4
Analyt- eller komponentnavne Ezetimibe Ketone Benzyl Ether
Forsendelsestilstand Room Temperature
Molekylær formel C31 H25 F2 N O3
Certificeringer/compliance ISO 17025
Synonym (3R,4S)-4-[4-(Benzyloxy)phenyl]-1-(4-fluorophenyl)-3-[3-(4-fluorophenyl)-3-oxopropyl]azetidin-2-one, 2-Azetidinone, 1-(4-fluorophenyl)-3-[3-(4-fluorophenyl)-3-oxopropyl]-4-[4-(phenylmethoxy)phenyl]-, (3R,4S)-, -Azetidinone, 1-(4-fluorophenyl)-3-[3-(4-fluorophenyl)-3-oxopropyl]-4-[4-(phenylmethoxy)phenyl]-, (3R-trans)-, Ezetimibe Ketone Methyl Ether
InChI formel InChI=1S/C31H25F2NO3/c32-24-10-6-22(7-11-24)29(35)19-18-28-30(34(31(28)36)26-14-12-25(33)13-15-26)23-8-16-27(17-9-23)37-20-21-4-2-1-3-5-21/h1-17,28,30H,18-20H2/t28-,30-/m1/s1
SMIL Fc1ccc(cc1)N2[C@@H]([C@@H](CCC(=O)c3ccc(F)cc3)C2=O)c4ccc(OCc5ccccc5)cc4
IUPAC navn (3R,4S)-1-(4-fluorophenyl)-3-[3-(4-fluorophenyl)-3-oxopropyl]-4-(4-phenylmethoxyphenyl)azetidin-2-one
Molekylvægt (g/mol) 497.53
Mængde 100 mg
Formel vægt 497.1803 g/mol