Få mere at vide
MedChemExpress Ezatiostat
Artikelnummer. 16448457
Encompass_Preferred
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
16448457 5 mg
1 options
Artikelnummer. 16448457 Leverandør MedChemExpress Leverandørnr. HY13634A
MedChemExpress Ezatiostat (TER199 free base; TLK199) is a tripeptide analog of glutathione and is a selective and orally active glutathione S-transferase P1-1 (GSTP1) inhibitor. Ezatiostat leads to JNK activation by inhibiting GSTP1. Ezatiostat stimulates both lymphocyte production and bone marrow progenitor proliferation. Ezatiostat has the potential for myelodysplastic syndrome (MDS) treatment.
Tekniske data
Renhed 98.0%
CAS 168682-53-9
Indhold og opbevaring Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
Til brug med (applikation) Cancer-Kinase/protease
Molekylvægt (g/mol) 529.65 g/mol
Produkttype Peptides
Mængde 5 mg
Forsendelsestilstand Room temperature
Opløselighed DMSO : ≥ 100 mg/mL (188.80 mM)
Synonym TER199(free base)∣ TLK199