Få mere at vide
EZ-Link NHS-PEG12-Biotin, TRC
Artikelnummer. 30475890
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30475890 250 mg
30482030 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30475890 Leverandør Toronto Research Chemicals Leverandørnr. TRCN394010250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 365441-71-0
Molekylær formel C19 H28 N4 O7 S
Synonym α-[3-[(2,5-Dioxo-1-pyrrolidinyl)oxy]-3-oxopropyl]-ω-[2-[[5-[(3aS,4S,6aR)-hexahydro-2-oxo-1H-thieno[3,4-d]imidazol-4-yl]-1-oxopentyl]amino]ethoxy]poly(oxy-1,2-ethanediyl)
Tekniske data
Certificeringer/compliance ISO 9001
Molekylvægt (g/mol) 456.513