Learn More
Memmert™ Extension of DAkkS Factory Calibration for UN160
Extension of DAkkS factory calibration by an additional freely selectable temperature for Memmert Ovens.
Brand:  Memmert™ UN160E39697
Product Code. 17549875
  • 7275.00 DKK / Each
Description
  • Extension of DAkkS calibration at Memmert factory by an additional freely selectable temperature value according to method C (DKD-R 5-7)
Specifications
Specifications
UN160 Oven
Factory Calibration Extension
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Memmert™ Extension of DAkkS Factory Calibration for UN160 > For Use With: UN160