Learn More
Memmert™ Extended Overtemperature Protection for UN160
Extended overtemperature protection with integrated Pt100 sensor for independent temperature monitoring.
Brand:  Memmert™ UN160A6
Product Code. 17549845
  • 1425.00 DKK / Each
Only null left
Estimated Shipment: 31-07-2025
Add to basket
This item is not returnable. View return policy
Description
Description
  • Accessory
  • With integrated Pt100 sensor
  • For models with SingleDISPLAY
Specifications
Specifications
UN160 Oven
Integrated Pt100 Sensor
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Extended Overtemperature Protection for UN160 > For Use With: UN160