Få mere at vide
Exemestane EP Impurity E (Exemestane USP Related Compound A), Mikromol™
Artikelnummer. 30462161
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30462161 4 x 25 mg
30050681 25 mg
17982436 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30462161 Leverandør Mikromol Leverandørnr. MM1161.0100254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Exemestane EP Impurity E (Exemestane USP Related Compound A), Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,2-Dihydro Exemestane
CAS 19457-55-7
Analyt- eller komponentnavne 1,2-Dihydro Exemestane
Forsendelsestilstand Room Temperature
Molekylær formel C20 H26 O2
Certificeringer/compliance ISO 17025
Synonym 6-Methylideneandrost-4-ene-3,17-dione, Exemestane Imp. E (EP), Exemestane USP Related Compound A, Exemestane USP RC A, Androst-4-ene-3,17-dione, 6-methylene-, 6-Methyleneandrost-4-ene-3,17-dione, 6-Methyleneandrost-4-en-3,17-dione, 6-Methylideneandrost-4-ene-3,17-dione, 1,2-Dihydroexemestane, Exemestane Related Compound A, Exemestane Impurity E
InChI formel InChI=1S/C20H26O2/c1-12-10-14-15-4-5-18(22)20(15,3)9-7-16(14)19(2)8-6-13(21)11-17(12)19/h11,14-16H,1,4-10H2,2-3H3/t14-,15-,16-,19+,20-/m0/s1
Løsningstype Neat
SMIL C[C@]12CC[C@H]3[C@@H](CC(=C)C4=CC(=O)CC[C@]34C)[C@@H]1CCC2=O
IUPAC navn (8R,9S,10R,13S,14S)-10,13-dimethyl-6-methylidene-1,2,7,8,9,11,12,14,15,16-decahydrocyclopenta[a]phenanthrene-3,17-dione
Molekylvægt (g/mol) 298.42
Mængde 4 x 25 mg
Formel vægt 298.1933 g/mol