missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ EWSR1/ATF1 DY Translocation FISH Probe
Labeled FISH probes for identification of gene translocation using Fluoresecent In Situ Hybridization Technique.
11735.00 DKK - 19325.00 DKK
Specifications
Content And Storage | Store at 4°C in the dark |
---|---|
Format | Tube |
For Use With (Application) | Fluorescent In Situ Hybridization (Cell) |
Gene | Ewing sarcoma breakpoint region 1 |
Gene Alias | EWS |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16043575
|
Abnova™
FT0022.100UL |
100 μL |
11735.00 DKK
100µL |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16053575
|
Abnova™
FT0022.200UL |
200 μL |
19325.00 DKK
200µL |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Store at 4°C in the dark | |
Fluorescent In Situ Hybridization (Cell) | |
EWS | |
2130;466 | |
Liquid |
Tube | |
Ewing sarcoma breakpoint region 1 | |
EWSR1 | |
DNA/RNA |