Få mere at vide
Invitrogen™ EVOS™ Light Cube, AO
:
For use with EVOS fl; AO (acridine orange, longpass emission)
Beskrivelse
- Ideal for use with EVOS FL Color Imaging System, EVOS FL Auto Imaging System or EVOS FL Imaging Systems
- Each cube contains an LED, condensing optics, and hard-coated filters
- Continuous illumination enables consistent results 50,000+ hour bulb lifetime that affords lower operating costs
- Adjustable light intensity that helps reduce photobleaching User changeable and automatically recognized by the system
Tekniske data
Tekniske data
| Type | AO Fluorescent Light Cube |
| Produktlinje | EVOS |
| Til brug med (udstyr) | Countess™ II FL Automated Cell Counter, EVOS™ M5000 Imaging System, EVOS™ M7000 Imaging System |
| Inkluderer | LED, Condensing Optics, Hard-coated Filters |
| Bølgelængde | Ex: 470/22 Em: 488/20 |
| Mængde | 1 Ea. |