Få mere at vide
Safety First Aid Evolution Plus Hydrofluoric Acid Burn Kit
Beskrivelse
- The kit is supplied in an Evolution case with a quick release wall mounting bracket, meaning the kit can be safely stored on the wall in a visible location and quickly removed to be taken to a casualty.
- Kit Contents:
- 4x HypaCover Low Adherent Absorbent Dressings, 10 x 10 cm
- 4x HypaBand Conforming Bandages, 7.5 cm x 4 m
- 4x HypaTouch Nitrile Gloves
- 2x C-gel Calcium Gluconate, 40 g
Tekniske data
Tekniske data
| Produkttype | First Aid Kit |
| Materiale | Plastic |
| Sterilitet | Sterile |