Få mere at vide
Safety First Aid Evolution Motorcycle First Aid Kit, BS8599-2 Compliant
Beskrivelse
- The first aid contents are supplied in a durable Evolution case, keeping them well protected.
- The compact case means the kit is easily portable.
- There is also a bold white cross on the green lid, making it easy to identify the contents.
- The BS8599-2 Motorcycle First Aid Kit in Case Contains:
- 1 x First Aid Guidance Leaflet
- 1 x Content List
- 1 x Trauma Dressing
- 5 x Cleansing Wipes
- 5 x Sterile Adhesive Dressings
- 1 x Nitrile Disposable Gloves
- 1 x Resuscitation Face Shield
- 1 x Burn Dressing
- 1 x Clothing Cutters
Tekniske data
Tekniske data
| Produkttype | First Aid Kit |
| Materiale | Plastic |
| Sterilitet | Sterile |