Få mere at vide
Thermo Scientific™ Evolution™ 260 Bio UV-Visible Spectrophotometer
Artikelnummer. p-4532038 Shop alle Thermo Scientific produkter
Type:
Evolution 260 Bio Computer Ctl
Inkluderer:
EU/US/UK strømkabler
Pakningsstørrelse:
Hver
Artikelnummer. 10438278
Brand: Thermo Scientific™ 840211000
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
The Thermo Scientific™ Evolution 260 spectrophotometer offers reliable, high-quality performance and enhanced customer usability.
New sales of this model have been discontinued. Please see the newest models: the Evolution Pro Spectrophotometers.
Complete Life Science Applications
Next generation INSIGHT software provides the streamlined convenience of easy-to-understand nucleic acid and protein analysis methods and includes dedicated modules for:
  • Nucleic Acid concentration by direct UV measurement at 260nm including DNA, ssDNA, RNA, and siRNA
  • Nucleic acid purity measurements from 260/280nm and 260/230 ratios
  • Protein analysis by direct UV measurement at 205 or 280nm
  • Most common colorimetric analysis methods including: Pierce 660nm Protein assay, Bradford, BCA and Lowry
  • Measuring the fluorescence labeling efficiency for proteins and nucleic acids
  • Measuring the microarray labeling efficiency with one or two dyes
  • Customizable, Straightforward INSIGHT Software
Personalize your home screen for easy navigation and convenience
  • Workbook tools save time and ensure accurate results
  • CUE scripting allows creation of customized interface and simplified workflow
Best-in-Class Performance
  • Double-beam geometry ideal for kinetics or any sample that may change over time
  • Application Focused Beam Geometry (AFBG) technology provides superior results for microcells and turbid samples
  • Xenon flash lamp requires no warm-up time allowing instant measurements
  • Electronic triggering provides the highest level of accuracy for rapid-mixing kinetics measurements
  • Precision monochromator drive delivers fast scanning data collection and wavelength accuracy Powerful Convenience at your Fingertips
  • Color, touch screen provides instrument control using built-in computer
  • Integrated keypad allows communication with INSIGHT software
  • Choice of Local or PC configurations provides flexibility and convenience
Accessories for all your Sampling Needs
  • Wide accessory selection with various temperature control options, sample changers, sippers and solid sampling devices complete your analysis
  • Automated cell changers with Peltier temperature control increase sample throughput
  • Choice of Peltier Single Cell Holder or a Smart 8-Cell Peltier system for temperature control and sample monitoring from 0° to 100°C
  • Easy software integration gives you complete control of your measurements
Reliable Assurance of System Performance
  • The Thermo Scientific UV Validator package provides support for system qualification and validation activities
  • Complete documentation facilitates compliance of your system with the requirements of FDA, GxP, ISO 9001:2000 and ISPE 2001 guidelines and regulations
  • CVC and Mercury Lamp accessory automated testing ensures accuracy of your data
  • Thermo Scientific INSIGHTsecurity software provides complete tools to meet 21 CFR Part 11 requirements
TRUSTED_SUSTAINABILITY
Tekniske data
Grundlinjeplanhed ±0.0010 A, 200 to 800 nm, 1.0 nm SBW, smoothing
Certificeringer/compliance ISO 9001:2008
Detektor type Dual Silicon Photodiodes
Dimensioner (L x B x H) 62.2 x 48.6 x 27.9 cm (24 x 19 x 11 in.)
Produktlinje Evolution 260 Bio
Spektral båndbredde Variable: 1.0 nm, 2.0 nm
Type Evolution 260 Bio Computer Ctl
Bølgelængdeområde 190 to 1100 nm
Drift <0.0005 A/hr., 500 nm, 1.0 nm SBW, 1 hr. warmup
Elektriske krav 100/240 V, 50/60 Hz selected automatically, 150 W maximum
Inkluderer EU/US/UK strømkabler
Lampens levetid 7 years typical
Støj 0A: <0.00015A
1A: <0.00025A
2A: <0.00080A
260nm, 1.0nm SBW, RMS
Optisk design AFBG microcell optimized, AFBG fiber optic optimized, AFBG materials optimized
Pharmacopoeia Compliance Testing (Guaranteed Performance Specifications) Resolution (toluene in Hexane): ≥1.8A Photometric Accuracy (60mg/L K2Cr2O7): ±0.010A Stray Light: ≤1%T at 198nm: KCI; ≤0.05%AT at 220nm: Nal, KI Wavelength Accuracy: ±0.5nm 541.9, 546.1nm Hg emission lines, ±0.8nm full range Wavelength Repeatability: ≤0.05nm, repetitive scanning of 546.1nm Hg emission line
Fotometrisk display -0.3 - 4.0A
Fotometrisk rækkevidde >3.5A
Fotometrisk gentagelighed ±0.0002 A
Processor Microsoft Windows 7, Windows 8
Scan Ordinat-tilstande Absorbance, % Transmittance, % Reflectance, Kubelka-Munk, log (1/R), log (Abs), Abs*Factor, Intensity
Scanningshastighed <1 to 6000 nm/min. (Variable)
garanti 3 Years
Bølgelængde nøjagtighed ±0.5 nm (541.9 nm mercury lines), ±0.8 nm (full range 190 to 1100 nm)
Bølgelængde datainterval 10, 5, 2, 1.0, 0.5, 0.2, 0.1 nm
Vægt (engelsk) 32 lb.
Vægt (metrisk) 14.4 kg