Learn More
Trajan™ eVol™ Syringe without Needle
eVol™ is digitally controlled positive displacement dispensing system that can be programmed to reproducibly and accurately perform a wide variety of liquid handling procedures.
Brand:  Trajan™ 2910023
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 12957586
  • 670.00 DKK / Each
Estimated Shipment: 26-06-2024
to see stock.
Description
Description
Product
Specifications
Specifications
Syringe
1/Ea.
50 μL