Få mere at vide
Tocris Bioscience™ Everolimus
mTOR inhibitor;immunosuppressant
Brand: Tocris Bioscience™ 6188/10
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Everolimus is a mTOR inhibitor. Inhibits tumor proliferation in vitro and in vivo. Inhibits VEGF-induced HUVEC proliferation in vitro, but does not affect tumor vascular permeability in mouse models. Immunosuppressant. Activates autophagy.Tekniske data
| mTOR Inhibitor, Immunosuppressant | |
| 159351-69-6 | |
| 958.22g/mol | |
| mTOR Inhibitors |
| mTOR | |
| C53H83NO14 | |
| Store at -20°C | |
| 10 mg |
Produkttitel
Ser du en mulighed for forbedring?
For Research Use Only