missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Eutech™ Refilling Reference Electrolytes

Eutech™ offers Reference Standards and Solutions for use with Eutech™ pH electrodes

Brand:  Thermo Scientific™ 01X211208

Product Code. 12331393

  • 892.29 DKK / Each

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The measure of a liquid’s acidity and alkalinity, pH is one of the most common parameter measured in a wide variety of industries ranging from water and wastewater treatment, chemical production, agriculture research and production, environmental monitoring, chemical and life sciences research, biochemical and pharmaceutical research, electronics production to food processing and other industrial applications.

Eutech™ offers a wide range of pH buffers, calibration, and reference solutions for use with pH meters and electrodes.

Features:

  • Buffers and calibration solutions packed in 480 ml durable plastic bottles – economical and convenient for laboratory use.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Saturated Potassium Chloride with Silver Chloride for single junction pH electrodes
Eutech pH Electrode
Refilling Reference Electrolyte
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number514-01 Certificate TypeCertificate of Analysis Date17/12/2024 Catalog Number01X211208
Lot Number424-01 Certificate TypeCertificate of Analysis Date21/10/2024 Catalog Number01X211208
Lot Number334-01 Certificate TypeCertificate of Analysis Date28/08/2024 Catalog Number01X211208
Lot Number274-01 Certificate TypeCertificate of Analysis Date08/07/2024 Catalog Number01X211208
Lot Number144-01 Certificate TypeCertificate of Analysis Date03/04/2024 Catalog Number01X211208
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Eutech™ Refilling Reference Electrolytes >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.