Learn More
Eutech™ Conductivity Calibration Sachets
Get quick and accurate conductivity measurements in the field, laboratories, and schools.
Brand: Thermo Scientific Eutech ECCON447BS
Description
Get quick, accurate, and convenient conductivity measurements with NIST_traceable Eutech™ Conductivity Calibration Sachets. Simply insert electrode into sachet pack, calibrate, rinse and discard sachet – no separate container needed.
Features:
- Conductivity Standards Sachet -20 ml
- NIST Traceable
Specifications
KCl Conductivity Sachets | |
Calibration Solution | |
Field, School, Laboratory |
447 ÎĽS/cm | |
Conductivity Standard |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
BO1A | Certificate of Analysis | 28/03/2024 | ECCON447BS |
CY1A0326 | Certificate of Analysis | 28/03/2024 | ECCON447BS |
CY1A | Certificate of Analysis | 07/03/2024 | ECCON447BS |
CY1A | Certificate of Analysis | 16/02/2024 | ECCON447BS |
AZ1A | Certificate of Analysis | 15/05/2023 | ECCON447BS |