Få mere at vide
Eugenol acetate 1000 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Acetyl Eugenol |
| CAS | 93-28-7 |
| Analyt- eller komponentnavne | Acetyl Eugenol |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C12 H14 O3 |
| Koncentration | 1000 μg/mL |
| Certificeringer/compliance | ISO 17034 |
| Synonym | Eugenol acetate (6CI), Phenol, 2-methoxy-4-(2-propenyl)-, acetate (9CI), Phenol, 4-allyl-2-methoxy-, acetate (7CI,8CI), 2-Methoxy-4-(2-propenyl)phenol acetate, 4-Allyl-2-methoxyphenyl acetate, Aceteugenol, Acetyleugenol, Eugenyl acetate, NSC 1242, O-Acetyleugenol, Phenol, 2-methoxy-4-(2-propen-1-yl)-, 1-acetate, Eugenol Imp. I (EP), Eugenol Impurity I |
| InChI formel | InChI=1S/C12H14O3/c1-4-5-10-6-7-11(15-9(2)13)12(8-10)14-3/h4,6-8H,1,5H2,2-3H3 |
| SMIL | COc1cc(CC=C)ccc1OC(=O)C |
| Vis mere |