Learn More
ETV6/Tel Antibody (SR1935), R&D Systems™
Product Code. 18613064 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18613064 50 μg 50µL
18699974 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18613064 Supplier Novus Biologicals Supplier No. NBP32235550ul
 Restricted Item
Forventet forsendelse: 04-12-2026
A Novus product, part of R&D Systems. Rabbit Monoclonal Antibody
ETV6/Tel Monoclonal specifically detects ETV6/Tel in Human samples. It is validated for Flow Cytometry∣Immunocytochemistry/Immunofluorescence∣Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Antigen ETV6/Tel
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone SR1935
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias ETS translocation variant 6, ets variant 6, ets variant gene 6 (TEL oncogene), ETS-related protein Tel1, Tel, TEL/ABL, TEL1, TELTEL1 oncogene, transcription factor ETV6
Host Species Rabbit
Immunogen A synthesized peptide derived from human ETV6/Tel (Uniprot #: P41212)
Purification Method Affinity purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 2120
Target Species Human
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus