missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ETV5/ERM Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP2-98876AF532
Additional Details : Weight : 0.00970kg
Description
ETV5/ERM Polyclonal specifically detects ETV5/ERM in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ETV5/ERM | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
ERMEts-related protein ERM, ETS translocation variant 5, ets variant 5, ets variant gene 5 (ets-related molecule), ets-related molecule | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human ETV5/ERM. | |
0.1 mL | |
Cancer | |
2119 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |