missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ETS2 Antibody (PCRP-ETS2-1D9), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-14209AF350
Additional Details : Weight : 0.00970kg
Description
ETS2 Monoclonal antibody specifically detects ETS2 in Human samples. It is validated for Flow Cytometry, ELISA, ImmunofluorescenceSpecifications
ETS2 | |
Monoclonal | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Antibody | |
IgG2b |
Flow Cytometry, ELISA, Immunofluorescence | |
PCRP-ETS2-1D9 | |
Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
ETS2IT1, human erythroblastosis virus oncogene homolog 210protein C-ets-2, oncogene ETS-2, v-ets avian erythroblastosis virus E2 oncogene homolog 2, v-ets avian erythroblastosis virus E26 oncogene homolog 2, v-ets erythroblastosis virus E26 oncogene homolog 2 (avian) | |
Recombinant full-length human ETS2 protein (Uniprot: P15036) | |
0.1 mL | |
Stem Cell Markers | |
2114 | |
Store at 4°C in the dark. | |
Purified |