Få mere at vide
Etretinate-d3, TRC
Artikelnummer. 30379216
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30379216 10 mg
30377146 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30379216 Leverandør Toronto Research Chemicals Leverandørnr. TRCE93800210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1185237-13-1
Molekylær formel C23H27D3O3
Molekylvægt (g/mol) 357.5
Synonym (all-E)-9-(4-Methoxy-d3-2,3,6-trimethylphenyl)-3,7-dimethyl-2,4,6,8-nonatetraenoic Acid Ethyl Ester, Ro 10-9359-d3, Tegison-d3, Tigason-d3, Tigasone-d3
SMIL CC(/C=C/C1=C(C)C(C)=C(OC([2H])([2H])[2H])C=C1C)=C\C=C\C(C)=C\C(OCC)=O
Tekniske data
Kemisk navn eller materiale Etretinate-d3
Mængde 10 mg
InChI formel InChI=1S/C23H30O3/c1-8-26-23(24)14-17(3)11-9-10-16(2)12-13-21-18(4)15-22(25-7)20(6)19(21)5/h9-15H,8H2,1-7H3/b11-9+,13-12+,16-10+,17-14+/i7D3