Få mere at vide
(±)-Etodolac-d3 (1-ethyl-2,2,2-d3), CDN
Artikelnummer. 30326257
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30326257 0.005 g
30315729 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30326257 Leverandør CDN Leverandørnr. CDND72960.005G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale rac Etodolac-d3
CAS 1276197-46-6
Molekylær formel C17 D3 H18 N O3
Mængde 0.005 g
Synonym Etodolac-D3 (1-ethyl-2,2,2-D3), (±)-Etodolac-d3 (1-ethyl-2,2,2-d3), rac Etodolac-d3, 8-Ethyl-1-(ethyl-2,2,2-d3)-1,3,4,9-tetrahydropyrano[3,4-b]indole-1-acetic acid, 2-[8-Ethyl-1-(2,2,2-trideuterioethyl)-4,9-dihydro-3 H-pyrano[3,4-b]indol-1-yl]acetic acid
InChI formel InChI=1 S/C17H21NO3/c1-3-11-6-5-7-12-13-8-9-21-17(4-2,10-14(19)20)16(13)18-15(11)12/h5-7,18 H,3-4,8-10H2,1-2H3,(H,19,20)/i2D3
SMIL [2 H]C([2 H])([2 H])CC1(CC(=O)O)OCCc2c1[nH]c3c(CC)cccc23
IUPAC navn 2-[8-ethyl-1-(2,2,2-trideuterioethyl)-4,9-dihydro-3 H-pyrano[3,4-b]indol-1-yl]acetic acid
Molekylvægt (g/mol) 290.372
Formel vægt 290.171 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity