missing translation for 'onlineSavingsMsg'
Få mere at vide

Etilefrin Hydrochloride, TRC

Artikelnummer. 30373687
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30373687 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30373687 Leverandør Toronto Research Chemicals Leverandørnr. TRCE93295050MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Etilefrine hydrochloride
CAS 943-17-9
Molekylær formel C10 H15 N O2 . Cl H
Mængde 50 mg
Synonym Benzenemethanol, α-[(ethylamino)methyl]-3-hydroxy-, hydrochloride (1:1), Benzenemethanol, α-[(ethylamino)methyl]-3-hydroxy-, hydrochloride (9CI), Benzyl alcohol, α-[(ethylamino)methyl]-m-hydroxy-, hydrochloride (6CI,8CI), (±)-Ethylphenylephrine hydrochloride, (±)-N-Ethylnorphenylephrine hydrochloride, 1-(3-Hydroxyphenyl)-2-ethylaminoethanol hydrochloride, Apocretin, Circupon, Effontil, Effortil, Effortilvet, Ethyl adrianol, Ethylephrine hydrochloride, Eti-Puren, Etilefrine hydrochloride, Funasol, Kertasin, Phetanol, Pulsamin, Tonus-Forte, Updormin, dl-1-(3-Hydroxyphenyl)-1-hydroxy-2-ethylaminoethane hydrochloride, dl-1-(3-Hydroxyphenyl)-2-ethylaminoethanol hydrochloride, dl-Effortil hydrochloride, dl-Ethylphenylephrine hydrochloride, dl-Etilefrin hydrochloride, dl-N-Ethylnorphenylephrine hydrochloride, α-[(Ethylamino)methyl]-m-hydroxybenzyl alcohol hydrochloride
InChI formel InChI=1S/C10H15NO2.ClH/c1-2-11-7-10(13)8-4-3-5-9(12)6-8;/h3-6,10-13H,2,7H2,1H3;1H
SMIL Cl.CCNCC(O)c1cccc(O)c1
IUPAC navn 3-[2-(ethylamino)-1-hydroxyethyl]phenol;hydrochloride
Molekylvægt (g/mol) 217.69
Formel vægt 217.087
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.