Få mere at vide
Ethylnarceine Hydrochloride, LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Ethylnarceine Hydrochloride |
| CAS | 12246-80-9 |
| Analyt- eller komponentnavne | Ethylnarceine Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C25 H31 N O8 . Cl H |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Benzoic acid, 6-[2-[6-[2-(dimethylamino)ethyl]-4-methoxy-1,3-benzodioxol-5-yl]acetyl]-2,3-dimethoxy-, ethyl ester, hydrochloride (1:1), Benzoic acid, 6-[[6-[2-(dimethylamino)ethyl]-4-methoxy-1,3-benzodioxol-5-yl]acetyl]-2,3-dimethoxy-, ethyl ester, hydrochloride (9CI), Narceine, ethyl-, hydrochloride (8CI), Ethyl narceinate hydrochloride, Ethylnarceine hydrochloride, Narcyl |
| InChI formel | InChI=1S/C25H31NO8.ClH/c1-7-32-25(28)21-16(8-9-19(29-4)23(21)31-6)18(27)13-17-15(10-11-26(2)3)12-20-24(22(17)30-5)34-14-33-20;/h8-9,12H,7,10-11,13-14H2,1-6H3;1H |
| Løsningstype | Neat |
| SMIL | Cl.CCOC(=O)c1c(OC)c(OC)ccc1C(=O)Cc2c(CCN(C)C)cc3OCOc3c2OC |
| Vis mere |