Få mere at vide
Ethylmalonic Acid, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Ethylmalonic Acid |
| CAS | 601-75-2 |
| Molekylær formel | C5 H8 O4 |
| Mængde | 5 g |
| Synonym | Ethyl-propanedioic Acid, 1,1-Propanedicarboxylic Acid, 2-Ethylpropanedioic Acid, Ethylmalonic Acid, NSC 96615, α-Carboxybutyric Acid |
| InChI formel | InChI=1S/C5H8O4/c1-2-3(4(6)7)5(8)9/h3H,2H2,1H3,(H,6,7)(H,8,9) |
| SMIL | CCC(C(=O)O)C(=O)O |
| IUPAC navn | 2-ethylpropanedioic acid |
| Molekylvægt (g/mol) | 132.12 |
| Formel vægt | 132.0423 |