Learn More
Ethyl thiooxamate, 95%, Thermo Scientific Chemicals
Catalog Number p-3736505
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
16982-21-1
C4H7NO2S
133.17
MFCD00074903
YMBMCMOZIGSBOA-UHFFFAOYSA-N
ethyl 2-amino-2-thioxoacetate, ethyl carbamothioylformate, ethyl thioxamate, ethyl thiooxamidate, ethyl amino thioxo acetate, ethyl thiooxamat, ethyl aminothioxoacetate, ethyl 2-thiooxamate, acetic acid, aminothioxo-, ethyl ester, thiooxamic acid ethyl ester
2733398
ethyl 2-amino-2-sulfanylideneacetate
CCOC(=O)C(=S)N
16982-21-1
C4H7NO2S
133.17
MFCD00074903
YMBMCMOZIGSBOA-UHFFFAOYSA-N
ethyl 2-amino-2-thioxoacetate, ethyl carbamothioylformate, ethyl thioxamate, ethyl thiooxamidate, ethyl amino thioxo acetate, ethyl thiooxamat, ethyl aminothioxoacetate, ethyl 2-thiooxamate, acetic acid, aminothioxo-, ethyl ester, thiooxamic acid ethyl ester
2733398
ethyl 2-amino-2-sulfanylideneacetate
CCOC(=O)C(=S)N
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 60.0°C to 66.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula H2NCSCO2C2H5
Quantity 1 g
Formula Weight 133.17
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material Ethyl thiooxamate