Learn More
Ethylenediaminetetraacetic Acid Dipotassium Salt Dihydrate, Sigma-Aldrich™
Product Code. 15870691
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15870691 100 g 100g
15860691 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15870691 Supplier Sigma Aldrich Supplier No. 8190400100
 Restricted Item
Forventet forsendelse: 26-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 25102-12-9
Specifications
Melting Point 272°C
MDL Number MFCD00150036
Quantity 100 g
Synonym Ethylenediaminetetraacetic acid dipotassium salt dihydrate, Ethylenedinitrilotetraacetic acid dipotassium salt, Titriplex™ Potassium salt, Ethylenediaminetetraacetic acid dipotassium salt dihydrate
Molecular Weight (g/mol) 404.45
Grade Research