Få mere at vide
Ethylenediaminetetraacetic Acid-d16, TRC
Artikelnummer. 30374963
Skift visning
Mængde:
2 mg
Artikelnummer. Mængde
30374963 2 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30374963 Leverandør Toronto Research Chemicals Leverandørnr. TRCE9177512MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Edetic Acid (D16)
CAS 203805-96-3
Molekylær formel C10 D16 N2 O8
Mængde 2 mg
Synonym EDTA (D16)
InChI formel InChI=1S/C10H16N2O8/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20)/i1D2,2D2,3D2,4D2,5D2,6D2/hD4
SMIL [2H]OC(=O)C([2H])([2H])N(C([2H])([2H])C(=O)O[2H])C([2H])([2H])C([2H])([2H])N(C([2H])([2H])C(=O)O[2H])C([2H])([2H])C(=O)O[2H]
IUPAC navn deuterio 2-[[2-[bis(1,1-dideuterio-2-deuteriooxy-2-oxo-ethyl)amino]-1,1,2,2-tetradeuterio-ethyl]-(1,1-dideuterio-2-deuteriooxy-2-oxo-ethyl)amino]-2,2-dideuterio-acetate
Molekylvægt (g/mol) 308.34
Formel vægt 308.1911