Få mere at vide
Ethylenediaminetetraacetic Acid-d16, CDN
Artikelnummer. 30308976
Skift visning
Mængde:
0.1 g
Pakningsstørrelse:
0.1g
Artikelnummer. Mængde unitSize
30308976 0.1 g 0.1g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30308976 Leverandør CDN Leverandørnr. CDND54060.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Edetic Acid (D16)
CAS 203805-96-3
Molekylær formel C10 D16 N2 O8
Mængde 0.1 g
Synonym EDTA (D16)
InChI formel InChI=1 S/C10H16N2O8/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20)/i1D2,2D2,3D2,4D2,5D2,6D2/hD4
SMIL [2 H]OC(=O)C([2 H])([2 H])N(C([2 H])([2 H])C(=O)O[2 H])C([2 H])([2 H])C([2 H])([2 H])N(C([2 H])([2 H])C(=O)O[2 H])C([2 H])([2 H])C(=O)O[2 H]
IUPAC navn deuterio 2-[[2-[bis(1,1-dideuterio-2-deuteriooxy-2-oxo-ethyl)amino]-1,1,2,2-tetradeuterio-ethyl]-(1,1-dideuterio-2-deuteriooxy-2-oxo-ethyl)amino]-2,2-dideuterio-acetate
Molekylvægt (g/mol) 308.34
Formel vægt 308.1911 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity