Få mere at vide
Ethylene Urea-d4, TRC
Artikelnummer. 30372397
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30372397 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30372397 Leverandør Toronto Research Chemicals Leverandørnr. TRCE917782100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ethyleneurea-d4
CAS 62740-68-5
Renhedsgrad noter GC
Molekylær formel C3 2H4 H2 N2 O
Mængde 100 mg
Synonym Ethylene Urea D4
InChI formel InChI=1S/C3H6N2O/c6-3-4-1-2-5-3/h1-2H2,(H2,4,5,6)/i1D2,2D2
SMIL [2H]C1([2H])NC(=O)NC1([2H])[2H]
IUPAC navn 4,4,5,5-tetradeuterioimidazolidin-2-one
Molekylvægt (g/mol) 90.12
Formel vægt 90.0731
Procent renhed >95