Få mere at vide
Ethylene Terephthalate Linear Trimer, TRC
Artikelnummer. 30136780
Skift visning
Mængde:
2.5 mg
10 mg
25 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30136780 25 mg -
30007040 10 mg -
30006985 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30136780 Leverandør Toronto Research Chemicals Leverandørnr. TRCE91771425MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 16033-73-1
Molekylær formel C32H30O14
Molekylvægt (g/mol) 638.57
Synonym Terephthalic Acid Ester with Ethylene Glycol, Bis(2-hydroxyethyl) O,O'-((terephthaloylbis(oxy))bis(ethane-2,1-diyl)) Diterephthalate, Ethylene Glycol Terephthalate (4:3), Terephthalic Acid 2-Hydroxyethyl Ester Trimol Ester 2-Hydroxyethyl Ester;
SMIL O=C(OCCO)C1=CC=C(C(OCCOC(C2=CC=C(C(OCCOC(C3=CC=C(C(OCCO)=O)C=C3)=O)=O)C=C2)=O)=O)C=C1
Tekniske data
Kemisk navn eller materiale Ethylene Terephthalate Linear Trimer
Renhedsgrad noter HPLC
Mængde 25 mg
InChI formel InChI=1S/C32H30O14/c33-13-15-41-27(35)21-1-5-23(6-2-21)29(37)43-17-19-45-31(39)25-9-11-26(12-10-25)32(40)46-20-18-44-30(38)24-7-3-22(4-8-24)28(36)42-16-14-34/h1-12,33-34H,13-20H2
Formel vægt 638.1636
Procent renhed >95%