Learn More
Ethylene glycol polymer average Mn 800, BLD Pharm™
Product Code. 30947489
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30947489 100 g 100g
30943877 500 g 500g
30946161 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30947489 Supplier BLD Pharm Supplier No. BD00813732100g
 Restricted Item
Forventet forsendelse: 30-11-2026
Average Mn 800
Specifications
CAS 25322-68-3
Molecular Formula (C2H4O)nH2O
MDL Number MFCD00081839
Quantity 100 g
SMILES NONE
Chemical Name or Material Ethylene glycol polymer average Mn 800