missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylene glycol ethyl methyl ether, 97%, stab. with 0.01% BHT, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
It is employed in the reaction of metal salts of thymine and uracil with tetra-acetyl-α-glucopyranosyl bromide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed in the reaction of metal salts of thymine and uracil with tetra-acetyl-α-glucopyranosyl bromide.
Solubility
Fully miscible in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It is employed in the reaction of metal salts of thymine and uracil with tetra-acetyl-α-glucopyranosyl bromide.
Solubility
Fully miscible in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Density | 0.85 |
Boiling Point | 100°C to 102°C |
Flash Point | 2°C (36°F) |
Linear Formula | CH3OCH2CH2OCH2CH3 |
Refractive Index | 1.387 |
Quantity | 10 g |
UN Number | UN3271 |
Solubility Information | Fully miscible in water. |
Formula Weight | 104.15 |
Percent Purity | 97% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'