Få mere at vide
Ethylene Glycol Dilaurate, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Ethylene Glycol Dilaurate |
| CAS | 624-04-4 |
| Molekylær formel | C26H50O4 |
| Mængde | 100 mg |
| Synonym | Ethane-1,2-diyl Didodecanoate, Ethylene Dilaurate, Ethylene Glycol Didodecanoate, Ethylene Laurate, Glycol Dilaurate, NSC 406565, Dodecanoic Acid, 1,2-ethanediyl ester, Lauric Acid, Ethylene Ester, Ethylene Glycol, Dilaurate, Emalex EG-di-L |
| InChI formel | InChI=1S/C26H50O4/c1-3-5-7-9-11-13-15-17-19-21-25(27)29-23-24-30-26(28)22-20-18-16-14-12-10-8-6-4-2/h3-24H2,1-2H3 |
| SMIL | CCCCCCCCCCCC(=O)OCCOC(=O)CCCCCCCCCCC |
| Molekylvægt (g/mol) | 426.67 |
| Formel vægt | 426.3709 |