missing translation for 'onlineSavingsMsg'
Få mere at vide

Ethylene Diamine Dihydrochloride-d4, TRC

Artikelnummer. 30379812
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30379812 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30379812 Leverandør Toronto Research Chemicals Leverandørnr. TRCE91771250MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Ethylenediamine-1,1,2,2-D4 Dihydrochloride
CAS 34334-71-9
Molekylær formel C2 2H4 H4 N2 . 2 Cl H
Mængde 50 mg
Synonym 1,2-Ethane-1,1,2,2-d4-diamine, dihydrochloride (9CI), Ethylene-d4-diamine, dihydrochloride (8CI), 1,2-Ethane-1,1,2,2-d4-diamine dihydrochloride, Ethylene-d4 diamine dihydrochloride, 1,2-Diamino-1,1,2,2-tetradeuteroethane dihydrochloride, Ethylenediamine-
InChI formel InChI=1S/C2H8N2.2ClH/c3-1-2-4;;/h1-4H2;2*1H/i1D2,2D2;;
SMIL Cl.Cl.[2H]C([2H])(N)C([2H])([2H])N
IUPAC navn 1,1,2,2-tetradeuterioethane-1,2-diamine;dihydrochloride
Molekylvægt (g/mol) 137.045
Formel vægt 136.047
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.