Få mere at vide
Ethylene-d4 Glycol, CDN
Artikelnummer. 30320282
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30320282 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320282 Leverandør CDN Leverandørnr. CDND03391G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ethylene-d4 Glycol
CAS 2219-51-4
Molekylær formel C2 2H4 H2 O2
Mængde 1 g
InChI formel InChI=1 S/C2H6O2/c3-1-2-4/h3-4 H,1-2H2/i1D2,2D2
SMIL [2 H]C([2 H])(O)C([2 H])([2 H])O
IUPAC navn 1,1,2,2-tetradeuterioethane-1,2-diol
Molekylvægt (g/mol) 66.09
Formel vægt 66.0619 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity