Få mere at vide
Ethylene-Beta-ionol-d3, TRC
Artikelnummer. 30378375
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30378375 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30378375 Leverandør Toronto Research Chemicals Leverandørnr. TRCE91754210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ethylene-beta-ionol-d3
CAS 77265-43-1
Molekylær formel C15H21D3O
Mængde 10 mg
Synonym (1E)-3-methyl-1-(2,6,6-trimethyl-1-cyclohexen-1-yl)-1,4-pentadien-3-ol-d3
InChI formel InChI=1S/C15H24O/c1-6-15(5,16)11-9-13-12(2)8-7-10-14(13,3)4/h6,9,11,16H,1,7-8,10H2,2-5H3/b11-9+/i5D3
SMIL [2H]C([2H])([2H])C(O)(C=C)\C=C\C1=C(C)CCCC1(C)C
IUPAC navn (1E)-3-(trideuteriomethyl)-1-(2,6,6-trimethylcyclohexen-1-yl)penta-1,4-dien-3-ol
Molekylvægt (g/mol) 223.37
Formel vægt 223.2015