missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ethyle4-chloro-2-(trifluoromethyl)pyrimidine-5-carboxylate, 97%, Maybridge™
597.86 DKK - 3438.77 DKK
Chemical Identifiers
CAS | 720-01-4 |
---|---|
Molecular Formula | C8H6ClF3N2O2 |
Molecular Weight (g/mol) | 254.593 |
MDL Number | MFCD00173897 |
InChI Key | DSULCDCBENGHAX-UHFFFAOYSA-N |
Synonym | ethyl 4-chloro-2-trifluoromethyl pyrimidine-5-carboxylate, 4-chloro-2-trifluoromethyl-pyrimidine-5-carboxylic acid ethyl ester, 5-pyrimidinecarboxylic acid, 4-chloro-2-trifluoromethyl-, ethyl ester, 4-chloro-2-trifluoromethyl-pyrimidine-5-carboxylicacidethylester, ethyl 2-trifluoromethyl-4-chloropyrimidine-5-carboxylate, ethyl 4-chloro-2-trifluoromethyl pyrimidine-5-car, ethyl 4-chloro-2-trifluoromethyl-pyrimidine-5-carboxylate, ethyl-4-chloro-2-trifluoromethyl pyrimidine-5-carboxylate, 5-pyrimidinecarboxylicacid, 4-chloro-2-trifluoromethyl-, ethyl ester |
PubChem CID | 2737166 |
IUPAC Name | ethyl 4-chloro-2-(trifluoromethyl)pyrimidine-5-carboxylate |
SMILES | CCOC(=O)C1=CN=C(N=C1Cl)C(F)(F)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10180942
|
Maybridge
SPB04750DA |
1g |
597.86 DKK
1g |
Estimated Shipment 19-03-2021 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10096724
|
Maybridge
SPB04750DE |
5g |
1549.00 DKK
5g |
Estimated Shipment 19-03-2021 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10031934
|
Maybridge
SPB04750EA |
10g |
3438.77 DKK
10g |
Estimated Shipment 19-03-2021 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Chemical Identifiers
720-01-4 | |
254.593 | |
DSULCDCBENGHAX-UHFFFAOYSA-N | |
2737166 | |
CCOC(=O)C1=CN=C(N=C1Cl)C(F)(F)F |
C8H6ClF3N2O2 | |
MFCD00173897 | |
ethyl 4-chloro-2-trifluoromethyl pyrimidine-5-carboxylate, 4-chloro-2-trifluoromethyl-pyrimidine-5-carboxylic acid ethyl ester, 5-pyrimidinecarboxylic acid, 4-chloro-2-trifluoromethyl-, ethyl ester, 4-chloro-2-trifluoromethyl-pyrimidine-5-carboxylicacidethylester, ethyl 2-trifluoromethyl-4-chloropyrimidine-5-carboxylate, ethyl 4-chloro-2-trifluoromethyl pyrimidine-5-car, ethyl 4-chloro-2-trifluoromethyl-pyrimidine-5-carboxylate, ethyl-4-chloro-2-trifluoromethyl pyrimidine-5-carboxylate, 5-pyrimidinecarboxylicacid, 4-chloro-2-trifluoromethyl-, ethyl ester | |
ethyl 4-chloro-2-(trifluoromethyl)pyrimidine-5-carboxylate |
Specifications
ethyle4-chloro-2-(trifluoromethyl)pyrimidine-5-carboxylate | |
1g | |
97.0 | |
C8H6ClF3N2O2 | |
ethyl 4-chloro-2-trifluoromethyl pyrimidine-5-carboxylate, 4-chloro-2-trifluoromethyl-pyrimidine-5-carboxylic acid ethyl ester, 5-pyrimidinecarboxylic acid, 4-chloro-2-trifluoromethyl-, ethyl ester, 4-chloro-2-trifluoromethyl-pyrimidine-5-carboxylicacidethylester, ethyl 2-trifluoromethyl-4-chloropyrimidine-5-carboxylate, ethyl 4-chloro-2-trifluoromethyl pyrimidine-5-car, ethyl 4-chloro-2-trifluoromethyl-pyrimidine-5-carboxylate, ethyl-4-chloro-2-trifluoromethyl pyrimidine-5-carboxylate, 5-pyrimidinecarboxylicacid, 4-chloro-2-trifluoromethyl-, ethyl ester | |
CCOC(=O)C1=CN=C(N=C1Cl)C(F)(F)F | |
254.593 | |
254.59 |
Amber glass bottle | |
720-01-4 | |
97% | |
MFCD00173897 | |
DSULCDCBENGHAX-UHFFFAOYSA-N | |
ethyl 4-chloro-2-(trifluoromethyl)pyrimidine-5-carboxylate | |
2737166 | |
97% |