missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-aminobenzoate, 99+%, Thermo Scientific Chemicals

Catalog Number p-3757806
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
582-33-2
C9H11NO2
165.19
MFCD00007794
ZMCBYSBVJIMENC-UHFFFAOYSA-N
3-aminobenzoic acid ethyl ester, m-ethoxycarbonylaniline, 3-ethoxycarbonyl aniline, ethyl m-aminobenzoate, benzoic acid, 3-amino-, ethyl ester, m-aminobenzoic acid, ethyl ester, m-aminobenzoate, metacaine, 3-amino-benzoic acid ethyl ester, benzoic acid, m-amino-, ethyl ester
11400
CHEBI:81494
ethyl 3-aminobenzoate
CCOC(=O)C1=CC(=CC=C1)N
This item is not returnable. View return policy
582-33-2
C9H11NO2
165.19
MFCD00007794
ZMCBYSBVJIMENC-UHFFFAOYSA-N
3-aminobenzoic acid ethyl ester, m-ethoxycarbonylaniline, 3-ethoxycarbonyl aniline, ethyl m-aminobenzoate, benzoic acid, 3-amino-, ethyl ester, m-aminobenzoic acid, ethyl ester, m-aminobenzoate, metacaine, 3-amino-benzoic acid ethyl ester, benzoic acid, m-amino-, ethyl ester
11400
CHEBI:81494
ethyl 3-aminobenzoate
CCOC(=O)C1=CC(=CC=C1)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Brown to Yellow
Density 1.1070g/mL
Boiling Point 172°C to 175°C (13.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula H2NC6H4CO2C2H5
Refractive Index 1.5595 to 1.5615
Quantity 5 g
Beilstein 14, 389
Specific Gravity 1.107
Solubility Information Solubility in water: slightly soluble. Other solubilities: very soluble in ethanol,diethyl ether,soluble in chloroform
Formula Weight 165.19
Percent Purity 99+%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material Ethyl 3-aminobenzoate
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl 3-aminobenzoate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'