missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ethyle2-(3-methoxyphenyl)acetate, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
50 g
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Linear Formula | CH3OC6H4CH2C(O)OCH2CH3 |
Quantity | 10 g |
Formula Weight | 194.23 |
Percent Purity | 97% |
Chemical Name or Material | Ethyl 2-(3-methoxyphenyl)acetate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'