Få mere at vide
Ethylbenzene-2,3,4,5,6-d5, CDN
Artikelnummer. 30328389
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30328389 1 g -
30313991 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30328389 Leverandør CDN Leverandørnr. CDND19031G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ethylbenzene-2,3,4,5,6-D5
CAS 20302-26-5
Molekylær formel C8 D5 H5
Mængde 1 g
Synonym Benzene-d5, ethyl- (8 CI, 9 CI), Ethylbenzene-d5, Ethylbenzene-2,3,4,5,6-d5, Ethylbenzene-d5 (ethyl-d5), Ethylbenzene-2,3,4,5,6-D5
InChI formel InChI=1 S/C8H10/c1-2-8-6-4-3-5-7-8/h3-7 H,2H2,1H3/i3D,4 D,5 D,6 D,7 D
SMIL [2 H]c1c([2 H])c([2 H])c(CC)c([2 H])c1[2 H]
IUPAC navn 1,2,3,4,5-pentadeuterio-6-ethylbenzene
Molekylvægt (g/mol) 111.1958
Formel vægt 111.1096 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity