Få mere at vide
Ethylamine-N,N-d2 (gas), CDN
Artikelnummer. 30319080
Skift visning
Mængde:
2.5 g
5 g
Pakningsstørrelse:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30319080 2.5 g 2.5g
30322414 5 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319080 Leverandør CDN Leverandørnr. CDND05522.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ethylamine-N,N-D2
CAS 5852-45-9
Molekylær formel C2 D2 H5 N
Mængde 2.5 g
Synonym Ethanamine-d2 (9 CI), Ethylamine-d2 (7 CI), Ethylamine-N,N-d2 (8 CI), N,N-Dideuterioethylamine, Ethylamine-N,N-D2
InChI formel InChI=1 S/C2H7N/c1-2-3/h2-3H2,1H3/i/hD2
SMIL [2 H]N([2 H])CC
IUPAC navn N,N-dideuterioethanamine
Molekylvægt (g/mol) 47.096
Formel vægt 47.0704 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity