missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (S)-(+)-4-cyano-3-hydroxybutyrate, 97%, ee 98+%, Thermo Scientific Chemicals

Catalog Number p-7048122
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
312745-91-8
C7H11NO3
157.17
MFCD00672878
LOQFROBMBSKWQY-UHFFFAOYNA-N
ethyl s-4-cyano-3-hydroxybutyrate, s-ethyl 4-cyano-3-hydroxybutanoate, ethyl 3s-4-cyano-3-hydroxybutanoate, s-ethyl-4-cyano-3-hydroxybutyrate, ethyl s-+-4-cyano-3-hydroxybutyrate, butanoic acid, 4-cyano-3-hydroxy-, ethyl ester, 3s, 4-cyano-3-hydroxy-butyric acid ethyl ester, pubchem21112
7020868
ethyl (3S)-4-cyano-3-hydroxybutanoate
CCOC(=O)CC(O)CC#N
This item is not returnable. View return policy
312745-91-8
C7H11NO3
157.17
MFCD00672878
LOQFROBMBSKWQY-UHFFFAOYNA-N
ethyl s-4-cyano-3-hydroxybutyrate, s-ethyl 4-cyano-3-hydroxybutanoate, ethyl 3s-4-cyano-3-hydroxybutanoate, s-ethyl-4-cyano-3-hydroxybutyrate, ethyl s-+-4-cyano-3-hydroxybutyrate, butanoic acid, 4-cyano-3-hydroxy-, ethyl ester, 3s, 4-cyano-3-hydroxy-butyric acid ethyl ester, pubchem21112
7020868
ethyl (3S)-4-cyano-3-hydroxybutanoate
CCOC(=O)CC(O)CC#N

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Not miscible or difficult to mix with water.

Notes
Store away from water/moisture. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Density 1.114
Boiling Point 270°C
Flash Point >110°C (230°F)
Refractive Index 1.448
Quantity 5 g
Solubility Information Not miscible or difficult to mix with water.
Optical Rotation +5° (Neat)
Formula Weight 157.17
Percent Purity 97%
Assay ee ≥98%
Chemical Name or Material Ethyl (S)-(+)-4-cyano-3-hydroxybutyrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'