missing translation for 'onlineSavingsMsg'
Få mere at vide

Ethyl Propionylacetate-d3, TRC

Artikelnummer. 30370037
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30370037 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30370037 Leverandør Toronto Research Chemicals Leverandørnr. TRCE9253971MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Ethyl Propionylacetate-d3
CAS 1346598-17-1
Molekylær formel C7H9D3O3
Mængde 1 mg
Synonym 3-Oxopentanoic Acid Ethyl Ester, 3-Oxovaleric Acid Ethyl Ester, 3-Oxopentanoic Acid Ethyl Ester, Ethyl 3-Ketopentanoate, Ethyl 3-Oxopentanoate, Ethyl 3-Oxovalerate, Ethyl Propanoylacetate, Ethyl Propioacetate, Ethyl .beta.-oxovalerate, NSC 7251, NSC 78662, Propionylacetic Acid Ethyl Ester
InChI formel InChI=1S/C7H12O3/c1-3-6(8)5-7(9)10-4-2/h3-5H2,1-2H3/i1D3
SMIL [2H]C([2H])([2H])CC(=O)CC(=O)OCC
IUPAC navn ethyl 5,5,5-trideuterio-3-oxopentanoate
Molekylvægt (g/mol) 147.19
Formel vægt 147.0975
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.